3-Amino-2,5-dimethylbenzonitrile 97%

Supplier: Ambeed
Warning

A274530-100MG A274530-1G A274530-250MG
77194-046EA 88.58 USD
77194-046 77194-048 77194-050
3-Amino-2,5-dimethylbenzonitrile 97%
3-Amino-2,5-dimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.18 g/mol
MDL Number: MFCD18826302
CAS Number: 90557-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR