1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea 97%

Supplier: Ambeed
Warning

A274522-1G A274522-5G
77194-042EA 31.93 USD
77194-042 77194-044
1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea 97%
1,3-Bis(benzyloxycarbonyl)-2-methyl-2-thiopseudourea

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₄S
MW: 358.41 g/mol
MDL Number: MFCD00674040
CAS Number: 25508-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR