(2-(Cyclopropylamino)pyrimidin-5-yl)boronic acid 98%

Supplier: Ambeed
Warning

A274429-100MG A274429-1G A274429-250MG A274429-5G
77166-532EA 44.29 USD
77166-532 77166-534 77166-536 77166-538
(2-(Cyclopropylamino)pyrimidin-5-yl)boronic acid 98%
(2-(Cyclopropylamino)pyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BN₃O₂
MW: 178.98 g/mol
MDL Number: MFCD18259229
CAS Number: 1312942-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR