2,5-Dimethyl-3-furancarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dimethyl-3-furoic Acid, 2,5-Dimethylfuran-3-carboxylic acid

A274416-1G A274416-25G A274416-5G
77166-526EA 27.81 USD
77166-526 77166-528 77166-530
2,5-Dimethyl-3-furancarboxylic acid 98%
2,5-Dimethyl-3-furancarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈O₃
MW: 140.14 g/mol
MDL Number: MFCD00082181
CAS Number: 636-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR