(1,3-Bis(2,6-diisopropylphenyl)imidazolidene) (3-chloropyridyl) palladium(II) dichloride 98%

Supplier: Ambeed
Warning

A274006-100MG A274006-1G A274006-250MG
77166-520EA 9 USD
77166-520 77166-522 77166-524
(1,3-Bis(2,6-diisopropylphenyl)imidazolidene) (3-chloropyridyl) palladium(II) dichloride 98%
(1,3-Bis(2,6-diisopropylphenyl)imidazolidene) (3-chloropyridyl) palladium(II) dichloride

Precaución: for Research Use Only

Formula: C₃₂H₄₂Cl₃N₃Pd
MDL Number: MFCD08457656
CAS Number: 927706-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR