5-Methoxy-1-methyl-1H-benzo[d]imidazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A273592-100MG A273592-1G A273592-250MG
77166-504EA 29 USD
77166-504 77166-506 77166-508
5-Methoxy-1-methyl-1H-benzo[d]imidazol-2(3H)-one 95%
5-Methoxy-1-methyl-1H-benzo[d]imidazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
MW: 178.18 g/mol
MDL Number: MFCD21362674
CAS Number: 4583-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR