6-Bromoimidazo[1,2-a]pyridin-8-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
8-Amino-6-bromoimidazo[1,2-a]pyridine
A273004-10G
A273004-1G
A273004-250MG
A273004-25G
A273004-5G
77166-488EA
185.4
USD
77166-488
77166-490
77166-492
77166-494
77166-496
6-Bromoimidazo[1,2-a]pyridin-8-amine 98%
6-Bromoimidazo[1,2-a]pyridin-8-amine
Precaución: for Research Use Only
Formula:
C₇H₆BrN₃ MW: 212.0500 g/mol |
MDL Number:
MFCD13250043 CAS Number: 676371-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...