tert-Butyldiphenylsilane 98%

Supplier: Ambeed
Warning

A272543-1G A272543-250MG A272543-5G
77166-474EA 26 USD
77166-474 77166-476 77550-690
tert-Butyldiphenylsilane 98%
tert-Butyldiphenylsilane

Precaución: for Research Use Only

Formula: C₁₆H₂₀Si
MDL Number: MFCD05663906
CAS Number: 33729-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR