5-Methoxy-3-oxopentanenitrile 95%

Supplier: Ambeed
Warning

A271282-100MG A271282-250MG
77166-464EA 104 USD
77166-464 77166-466
5-Methoxy-3-oxopentanenitrile 95%
5-Methoxy-3-oxopentanenitrile

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.14 g/mol
MDL Number: MFCD11102787
CAS Number: 97820-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR