2-(2-Chloroethyl)thiophene 98%

Supplier: Ambeed
Danger

A271185-25G A271185-5G
77166-460EA 80.34 USD
77166-460 77166-462
2-(2-Chloroethyl)thiophene 98%
2-(2-Chloroethyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₇ClS
MDL Number: MFCD11646274
CAS Number: 19995-38-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR