6-Amino-3-methylpyrimidin-4(3H)-one 95%

Supplier: Ambeed
Danger

A271159-100MG A271159-250MG A271159-1G
77166-456EA 244.01 USD
77166-456 77166-458 77892-524
6-Amino-3-methylpyrimidin-4(3H)-one 95%
6-Amino-3-methylpyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MDL Number: MFCD19204434
CAS Number: 1122-46-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR