6-Amino-5-(prop-1-yn-1-yl)pyrimidin-2(1H)-one 97%

Supplier: Ambeed
Warning

A271086-100MG A271086-250MG
77194-026EA 86.52 USD
77194-026 77194-028
6-Amino-5-(prop-1-yn-1-yl)pyrimidin-2(1H)-one 97%
6-Amino-5-(prop-1-yn-1-yl)pyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₇N₃O
MW: 149.15 g/mol
MDL Number: MFCD19203699
CAS Number: 151091-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR