6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A271041-100MG A271041-250MG
77166-452EA 79.31 USD
77166-452 77166-454
6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 98%
6-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀FN·ClH
MW: 187.642 g/mol
MDL Number: MFCD08544167
CAS Number: 1191908-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR