2-Chloro-4-methoxy-7-methylbenzo[d]thiazole 97%

Supplier: Ambeed
Danger

A271008-100MG A271008-1G A271008-250MG A271008-5G
77194-014EA 57 USD
77194-014 77194-016 77194-018 77194-020
2-Chloro-4-methoxy-7-methylbenzo[d]thiazole 97%
2-Chloro-4-methoxy-7-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₉H₈ClNOS
MDL Number: MFCD04971832
CAS Number: 108773-00-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR