5-Bromo-3-chloro-2-fluorobenzotrifluoride 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-chloro-2-fluoro-3-(trifluoromethyl)benzene

A270919-1G A270919-25G
77166-448EA 39.14 USD
77166-448 77166-450
5-Bromo-3-chloro-2-fluorobenzotrifluoride 95%
5-Bromo-3-chloro-2-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrClF₄
MW: 277.441 g/mol
MDL Number: MFCD09033157
CAS Number: 914225-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR