Boc-D-Arg(Pbf)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nα-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-D-arginine
A270915-1G
A270915-5G
A270915-250MG
A270915-25G
77166-446EA
22.66
USD
77166-446
77593-674
77683-824
77683-825
Boc-D-Arg(Pbf)-OH 95%
Boc-D-Arg(Pbf)-OH
Precaución: For research use only.
Formula:
C₂₄H₃₈N₄O₇S MW: 526.649 g/mol |
MDL Number:
MFCD00236822 CAS Number: 186698-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...