4,6-Dichloropyridin-3-amine 97%

Supplier: Ambeed
Warning

A270767-100MG A270767-10G A270767-1G A270767-250MG A270767-25G A270767-5G
77166-426EA 6 USD
77166-426 77166-428 77166-430 77166-432 77166-434 77166-436
4,6-Dichloropyridin-3-amine 97%
4,6-Dichloropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD00234054
CAS Number: 7321-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR