1-Methyl-1H-pyrazole-4-carboxamide 97%

Supplier: Ambeed
Warning

A270675-100MG A270675-1G A270675-250MG A270675-5G
77194-002EA 25.75 USD
77194-002 77194-004 77194-006 77194-008
1-Methyl-1H-pyrazole-4-carboxamide 97%
1-Methyl-1H-pyrazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD11643542
CAS Number: 89280-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR