Tetrakis(dimethylsiloxy)silane 96%

Supplier: Ambeed
Warning

A270572-100G A270572-25G A270572-5G
77166-416EA 60 USD
77166-416 77166-418 77166-420
Tetrakis(dimethylsiloxy)silane 96%
Tetrakis(dimethylsiloxy)silane

Precaución: for Research Use Only

Formula: C₈H₂₈O₄Si₅
MDL Number: MFCD00053848
CAS Number: 17082-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR