2-Bromo-N,4-dimethylaniline 95%

Supplier: Ambeed
Warning

A270287-100MG A270287-250MG
77166-410EA 23.69 USD
77166-410 77166-412
2-Bromo-N,4-dimethylaniline 95%
2-Bromo-N,4-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD11150924
CAS Number: 81090-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR