1,2,3,5-Tetramethylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: Isodurene

A270145-1G A270145-5G A270145-25G
77193-996EA 22.66 USD
77193-996 77193-998 77746-475
1,2,3,5-Tetramethylbenzene 98%
1,2,3,5-Tetramethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄
MW: 134.22 g/mol
MDL Number: MFCD00008524
CAS Number: 527-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR