2-(Methyl(2-phenoxyethyl)amino)ethanol 95%

Supplier: Ambeed
Warning

A270087-100MG A270087-1G A270087-250MG
77166-404EA 12.36 USD
77166-404 77166-406 77166-408
2-(Methyl(2-phenoxyethyl)amino)ethanol 95%
2-(Methyl(2-phenoxyethyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
MDL Number: MFCD16152087
CAS Number: 113824-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR