Ethyl 4-chloro-6-methylpyrimidine-5-carboxylate 97%

Supplier: Ambeed
Danger

A269684-250MG A269684-100MG
77166-390EA 94 USD
77166-390 77892-519
Ethyl 4-chloro-6-methylpyrimidine-5-carboxylate 97%
Ethyl 4-chloro-6-methylpyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MDL Number: MFCD25959552
CAS Number: 157981-60-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR