(R)-2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile 97%
Supplier: Ambeed
|
Warning
|
A269523-100MG
A269523-1G
A269523-250MG
77166-384EA
71
USD
77166-384
77166-386
77166-388
(R)-2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile 97%
(R)-2-(9-(Pyridin-2-yl)-6-oxaspiro[4.5]decan-9-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₁₆H₂₀N₂O |
MDL Number:
MFCD28134617 CAS Number: 1401031-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...