4,4-Dimethyl-1,4-azasilinane hydrochloride 95%

Supplier: Ambeed
Warning

A269157-100MG A269157-1G A269157-250MG
77166-376EA 79.31 USD
77166-376 77166-378 77166-380
4,4-Dimethyl-1,4-azasilinane hydrochloride 95%
4,4-Dimethyl-1,4-azasilinane hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆ClNSi
MW: 165.73 g/mol
MDL Number: MFCD28399395
CAS Number: 130596-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR