Decyl 4-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A268584-1G A268584-5G
77166-372EA 33.99 USD
77166-372 77166-374
Decyl 4-hydroxybenzoate 95%
Decyl 4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₇H₂₆O₃
MW: 278.39 g/mol
MDL Number: MFCD01661520
CAS Number: 69679-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR