10-Phenyldecanoic acid 97%

Supplier: Ambeed
Warning

A268554-100MG A268554-250MG
77193-984EA 30 USD
77193-984 77193-986
10-Phenyldecanoic acid 97%
10-Phenyldecanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₄O₂
MDL Number: MFCD00037804
CAS Number: 18017-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR