2-(5-Methoxypyridin-2-yl)acetonitrile 95%

Supplier: Ambeed
Warning

A268265-100MG A268265-250MG A268265-1G
77166-370EA 47 USD
77166-370 77892-516 77892-515
2-(5-Methoxypyridin-2-yl)acetonitrile 95%
2-(5-Methoxypyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD12028722
CAS Number: 204067-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR