4-(4-Tetrahydropyranyl)phenylboronic Acid Pinacol Ester 95%
Supplier: Ambeed
|
Warning
|
A268097-1G
A268097-250MG
A268097-5G
77166-362EA
199
USD
77166-362
77166-364
77166-366
4-(4-Tetrahydropyranyl)phenylboronic Acid Pinacol Ester 95%
4-(4-Tetrahydropyranyl)phenylboronic Acid Pinacol Ester
Precaución: for Research Use Only
Formula:
C₁₇H₂₅BO₃ |
MDL Number:
MFCD18733758 CAS Number: 1312479-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...