4-(Indolin-5-yl)morpholine hydrochloride 95%

Supplier: Ambeed
Warning

A268044-100MG A268044-250MG
77166-358EA 15.45 USD
77166-358 77166-360
4-(Indolin-5-yl)morpholine hydrochloride 95%
4-(Indolin-5-yl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClN₂O
MW: 240.73 g/mol
MDL Number: MFCD28369478
CAS Number: 1646152-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR