2-Amino-3-bromo-5-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-cyano-4-fluoroaniline, 3-Bromo-5-fluoroanthranilonitrile

A267327-1G A267327-250MG
77193-968EA 144.2 USD
77193-968 77193-970
2-Amino-3-bromo-5-fluorobenzonitrile 97%
2-Amino-3-bromo-5-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
MDL Number: MFCD22566641
CAS Number: 1403667-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR