3-Acetylpyridine 98%

Supplier: Ambeed
Danger

A267279-10G A267279-25G
77166-346EA 5 USD
77166-346 77166-348
3-Acetylpyridine 98%
3-Acetylpyridine

Precaución: for Research Use Only

Formula: C₇H₇NO
MDL Number: MFCD00006396
CAS Number: 350-03-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR