6,8-Dihydro-5H-imidazo[2,1-c][1,4]oxazine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A267189-1G A267189-250MG
77193-964EA 744.69 USD
77193-964 77193-966
6,8-Dihydro-5H-imidazo[2,1-c][1,4]oxazine-3-carbaldehyde 95%
6,8-Dihydro-5H-imidazo[2,1-c][1,4]oxazine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD18823007
CAS Number: 623564-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR