1,5-Diazobicyclo[4.3.0]-5-nonene 98%

Supplier: Ambeed
Danger

Synonyms: DBN, 2,3,4,6,7,8-Hexahydropyrrolo[1,2-a]pyrimidine, 1,5-Diazabicyclo[4.3.0]-5-nonene

A266941-100G A266941-10G A266941-25G
77193-958EA 40.17 USD
77193-958 77193-960 77193-962
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
1,5-Diazobicyclo[4.3.0]-5-nonene

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.19 g/mol
MDL Number: MFCD00005554
CAS Number: 3001-72-7
UN: 3267
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR