1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%

Supplier: Ambeed
Warning

Synonyms: Cyclohexane-1,3-dicarboxylic acid, 1,3-Cyclohexanedicarboxylic acid

A266918-100G A266918-1G A266918-5G
77193-950EA 78 USD
77193-950 77193-952 77193-954 77193-956
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 97%
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MDL Number: MFCD00134411
CAS Number: 3971-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR