Diallyl phthalate 98%

Supplier: Ambeed
Warning

Synonyms: Phthalic acid diallyl ester

A266629-1000G A266629-25G A266629-500G
77193-944EA 30.9 USD
77193-944 77193-946 77193-948
Diallyl phthalate 98%
Diallyl phthalate

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₄
MW: 246.26 g/mol
MDL Number: MFCD00008646
CAS Number: 131-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR