(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone 97%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide

A266487-10G A266487-1G A266487-250MG A266487-5G
77193-938EA 125.66 USD
77193-938 77193-940 77193-942 77571-654
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone 97%
(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₂O₂
MW: 384.470 g/mol
MDL Number: MFCD00145260
CAS Number: 96293-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR