7,8-Dihydro-2,5(1H,6H)-quinolinedione 98%

Supplier: Ambeed
Warning

A266017-1G A266017-250MG A266017-5G
77193-932EA 25.75 USD
77193-932 77193-934 77193-936
7,8-Dihydro-2,5(1H,6H)-quinolinedione 98%
7,8-Dihydro-2,5(1H,6H)-quinolinedione

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
MDL Number: MFCD00766821
CAS Number: 15450-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR