6-Chloro-2-fluoro-3-methylaniline 97%

Supplier: Ambeed
Danger

A264984-1G A264984-250MG
77193-928EA 228 USD
77193-928 77193-930
6-Chloro-2-fluoro-3-methylaniline 97%
6-Chloro-2-fluoro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇ClFN
MDL Number: MFCD06660187
CAS Number: 702640-48-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR