6-Nitro-4-quinazolone 95%

Supplier: Ambeed
Warning

Synonyms: 6-Nitroquinazolin-4(3H)-one, 6-Nitro-4(3H)-quinazolinone

A264952-100MG A264952-10G A264952-1G A264952-250MG A264952-25G A264952-5G
77166-308EA 6.18 USD
77166-308 77166-310 77166-312 77166-314 77166-316 77166-318
6-Nitro-4-quinazolone 95%
6-Nitro-4-quinazolone

Precaución: for Research Use Only

Formula: C₈H₅N₃O₃
MW: 191.1436 g/mol
MDL Number: MFCD00137845
CAS Number: 6943-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR