Sodium 3-[(3,5-Dimethoxyphenyl)(ethyl)amino]-2-hydroxypropane-1-sulfonate 95%

Supplier: Ambeed
Warning

A264792-100MG A264792-10MG A264792-1G A264792-1MG A264792-250MG
77166-298EA 37.08 USD
77166-298 77166-300 77166-302 77166-304 77166-306
Sodium 3-[(3,5-Dimethoxyphenyl)(ethyl)amino]-2-hydroxypropane-1-sulfonate 95%
Sodium 3-[(3,5-Dimethoxyphenyl)(ethyl)amino]-2-hydroxypropane-1-sulfonate

Precaución: for Research Use Only

Formula: C₁₃H₂₀NNaO₆S
MW: 341.35 g/mol
MDL Number: MFCD00057627
CAS Number: 83777-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR