4-(4-Chlorophenoxy)benzonitrile 98%

Supplier: Ambeed
Warning

A264712-1G A264712-250MG A264712-25G A264712-5G
77166-290EA 50 USD
77166-290 77166-292 77166-294 77166-296
4-(4-Chlorophenoxy)benzonitrile 98%
4-(4-Chlorophenoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈ClNO
MDL Number: MFCD08276330
CAS Number: 74448-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR