4-(1-Hydroxyethyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-alpha-methylbenzyl alcohol

A264671-100MG A264671-1G A264671-250MG A264671-5G
77193-916EA 11 USD
77193-916 77193-918 77193-920 77193-922
4-(1-Hydroxyethyl)phenol 97%
4-(1-Hydroxyethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MDL Number: MFCD00016858
CAS Number: 2380-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR