4-Methyl-6-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

A264444-100MG A264444-1G A264444-250MG A264444-5G
77166-284EA 17 USD
77166-284 77166-286 77166-288 77892-506
4-Methyl-6-nitro-1H-indazole 98%
4-Methyl-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD07781465
CAS Number: 885520-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR