2-Bromo-4-chloro-3-methylpyridine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A264156-250MG A264156-1G A264156-100MG
77166-280EA 21 USD
77166-280 77683-804 77892-505
2-Bromo-4-chloro-3-methylpyridine 95%, Ambeed.Inc
2-Bromo-4-chloro-3-methylpyridine

Precaución: For research use only.

Formula: C₆H₅BrClN
MDL Number: MFCD16609890
CAS Number: 1211521-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR