1-(5-Bromo-4-methylpyridin-2-yl)ethanone 98+%

Supplier: Ambeed
Warning

A264055-1G A264055-5G
77166-266EA 33.99 USD
77166-266 77166-268
1-(5-Bromo-4-methylpyridin-2-yl)ethanone 98+%
1-(5-Bromo-4-methylpyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MW: 214.05 g/mol
MDL Number: MFCD27981362
CAS Number: 1413285-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR