4',5'-Dimethoxy-2'-nitroacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4,5-Dimethoxy-2-nitrophenyl)ethanone

A263700-250MG A263700-5G
77193-902EA 16 USD
77193-902 77193-904
4',5'-Dimethoxy-2'-nitroacetophenone 95%
4',5'-Dimethoxy-2'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₅
MW: 225.20 g/mol
MDL Number: MFCD00130048
CAS Number: 4101-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR