1-Boc-3-(methylamino)azetidine 97%

Supplier: Ambeed
Danger

A263666-5G A263666-1G
77166-264EA 27 USD
77166-264 77612-522
1-Boc-3-(methylamino)azetidine 97%
1-Boc-3-(methylamino)azetidine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD08752496
CAS Number: 454703-20-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR