(2R,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A263644-100MG A263644-10G A263644-1G A263644-250MG A263644-25G A263644-5G
77166-254EA 7 USD
77166-254 77166-256 77166-258 77166-260 77166-262 77892-502
(2R,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid 95%
(2R,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₄
MDL Number: MFCD02684479
CAS Number: 681128-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR