Ethyl 5-chloro-3-formyl-1H-indole-2-carboxylate 96%
Supplier: Ambeed
|
Warning
|
A263468-1G
A263468-250MG
A263468-5G
77166-248EA
161.71
USD
77166-248
77166-250
77166-252
Ethyl 5-chloro-3-formyl-1H-indole-2-carboxylate 96%
Ethyl 5-chloro-3-formyl-1H-indole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀ClNO₃ MW: 251.66 g/mol |
MDL Number:
MFCD02257705 CAS Number: 43142-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...